accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1324-80-7

HNNHNHSOOOHONa

CAS RN 1324-80-7

IUPAC Name: sodium 2-[(4-{hydroxybis[4-(phenylamino)phenyl]methyl}phenyl)amino]benzene-1-sulfonate
Synonyms: sodium 2-[(4-{hydroxybis[4-(phenylamino)phenyl]methyl}phenyl)amino]benzenesulfonate
Molecular Weight (g/mol): 635.71
Molecular Formula: C37H30N3NaO4S
InChi Key: BPCZPTGXEYXQRS-UHFFFAOYSA-M
SMILES: [Na+].OC(C1=CC=C(NC2=CC=CC=C2)C=C1)(C1=CC=C(NC2=CC=CC=C2)C=C1)C1=CC=C(NC2=CC=CC=C2S([O-])(=O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.