accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 133-91-5

HOOIIHO

CAS RN 133-91-5

IUPAC Name: 2-hydroxy-3,5-diiodobenzoic acid
Synonyms: 3,5-diiodosalicylic acid
Molecular Weight (g/mol): 389.92
Molecular Formula: C7H4I2O3
InChi Key: DHZVWQPHNWDCFS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(I)=CC(I)=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.