accessibility menu, dialog, popup

UserName

Save up to 60% on cold storage essentials

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 141430-65-1

H3COSOOHNNNHHO

CAS RN 141430-65-1

IUPAC Name: N-{2-[(4-hydroxyphenyl)amino]pyridin-3-yl}-4-methoxybenzene-1-sulfonamide
Synonyms: N-{2-[(4-hydroxyphenyl)amino]pyridin-3-yl}-4-methoxybenzenesulfonamide
Molecular Weight (g/mol): 371.41
Molecular Formula: C18H17N3O4S
InChi Key: URCVCIZFVQDVPM-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CN=C1NC1=CC=C(O)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.