accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1610-17-9

CH3NHNNHCH3H3CNOH3CN

CAS RN 1610-17-9

IUPAC Name: N2-ethyl-6-methoxy-N4-(propan-2-yl)-1,3,5-triazine-2,4-diamine
Synonyms: primatol atratone
Molecular Weight (g/mol): 211.27
Molecular Formula: C9H17N5O
InChi Key: PXWUKZGIHQRDHL-UHFFFAOYSA-N
SMILES: CCNC1=NC(NC(C)C)=NC(OC)=N1
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.