accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 16642-92-5

OOFFF(E)

CAS RN 16642-92-5

IUPAC Name: (2E)-3-[4-(trifluoromethyl)phenyl]prop-2-enoate
Synonyms: (2E)-3-[4-(trifluoromethyl)phenyl]prop-2-enoate
Molecular Weight (g/mol): 215.15
Molecular Formula: C10H6F3O2
InChi Key: ANRMAUMHJREENI-ZZXKWVIFSA-M
SMILES: [O-]C(=O)\C=C\C1=CC=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.