accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1666-13-3

SeSe

CAS RN 1666-13-3

IUPAC Name: (phenyldiselanyl)benzene
Synonyms: diphenyl diselenide
Molecular Weight (g/mol): 312.15
Molecular Formula: C12H10Se2
InChi Key: YWWZCHLUQSHMCL-UHFFFAOYSA-N
SMILES: [Se]([Se]C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.