accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1731-92-6

H3COOCH3

CAS RN 1731-92-6

IUPAC Name: methyl heptadecanoate
Synonyms: heptadecanoic acid, methyl ester
Molecular Weight (g/mol): 284.48
Molecular Formula: C18H36O2
InChi Key: HUEBIMLTDXKIPR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC(=O)OC
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.