accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 17354-14-2

H3CHNHNCH3OO

CAS RN 17354-14-2

IUPAC Name: 1,4-bis(butylamino)-9,10-dihydroanthracene-9,10-dione
Synonyms: oil blue 35 colouring agent CI 61554 (solvent blue 35)
Molecular Weight (g/mol): 350.46
Molecular Formula: C22H26N2O2
InChi Key: OCQDPIXQTSYZJL-UHFFFAOYSA-N
SMILES: CCCCNC1=CC=C(NCCCC)C2=C1C(=O)C1=CC=CC=C1C2=O
1x1 image pixel for data collection 1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.