accessibility menu, dialog, popup

UserName

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 30379-55-6

HOOO

CAS RN 30379-55-6

IUPAC Name: 2-(benzyloxy)acetic acid
Synonyms: (benzyloxy)acetic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: GRZHHTYDZVRPIC-UHFFFAOYSA-N
SMILES: OC(=O)COCC1=CC=CC=C1
Boiling Point 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.