accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 523-87-5

H3CNCH3OCH3NONCH3NClHNO

CAS RN 523-87-5

IUPAC Name: 8-chloro-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione; [2-(diphenylmethoxy)ethyl]dimethylamine
Synonyms: travelin
Molecular Weight (g/mol): 469.97
Molecular Formula: C24H28ClN5O3
InChi Key: NFLLKCVHYJRNRH-UHFFFAOYSA-N
SMILES: CN1C2=C(NC(Cl)=N2)C(=O)N(C)C1=O.CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.