accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 531-95-3

HO(S)OOH

CAS RN 531-95-3

IUPAC Name: (3S)-3-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-7-ol
Synonyms: (-)-equol 4',7-isoflavandiol (S)-equol (S)-(-)-4',7-isoflavandiol 6'-hydroxy-equol 3'-hydroxy-equol 4',7-dihydroxyisoflavane (+-)-isomer of equol 4',7-dihydroxyisoflavan (+-)-of equol
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: ADFCQWZHKCXPAJ-GFCCVEGCSA-N
SMILES: OC1=CC=C(C=C1)[C@H]1COC2=CC(O)=CC=C2C1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.