accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 556-08-1

CH3ONHOHO

CAS RN 556-08-1

IUPAC Name: 4-acetamidobenzoic acid
Synonyms: 4-acetamidobenzoic acid N-acetyl-p-aminobenzoic acid acetyl-p-aminobenzoic acid P-carboxyacetanilide acedoben acedoben [inn-spanish] PAAB acedobene [inn-french] acedobene PAcBA
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: QCXJEYYXVJIFCE-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.