accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 558-17-8

CH3CH3CH3I

CAS RN 558-17-8

IUPAC Name: 2-iodo-2-methylpropane
Synonyms: propane, 2-iodo-2-methyl-
Molecular Weight (g/mol): 184.02
Molecular Formula: C4H9I
InChi Key: ANGGPYSFTXVERY-UHFFFAOYSA-N
SMILES: CC(C)(C)I
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.