accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 56718-71-9

H3COOH

CAS RN 56718-71-9

IUPAC Name: 4-(2-methoxyethyl)phenol
Synonyms: 4-(2-methoxyethyl)phenol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: FAYGEALAEQKPDI-UHFFFAOYSA-N
SMILES: COCCC1=CC=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.