accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 57842-10-1

CAS RN 57842-10-1

IUPAC Name: 3-iodopyridine-4-carboxylic acid
Synonyms: 3-iodopyridine-4-carboxylic acid
Molecular Weight (g/mol): 249.01
Molecular Formula: C6H4INO2
InChi Key: MJSNRTLYWDXHAQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(I)C=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.