accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 593-39-5

H3COHO(Z)

CAS RN 593-39-5

IUPAC Name: (6Z)-octadec-6-enoic acid
Synonyms: petroselinic acid cis-petroselinic acid cis-ω-12-octadecenoic acid delta-5-octadecylenic acid octadec-6c-ensaere petroselinsaeure petroselic acid petroseleninsaeure 6-octadecylenic acid (6Z)-petroselinic acid
Molecular Weight (g/mol): 282.47
Molecular Formula: C18H34O2
InChi Key: CNVZJPUDSLNTQU-SEYXRHQNSA-N
SMILES: CCCCCCCCCCC\C=C/CCCCC(O)=O
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.