accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 6027-91-4

CAS RN 6027-91-4

IUPAC Name: 1,2,5,6-tetrahydropyridine-3-carboxylic acid hydrochloride
Synonyms: guvacine hydrochloride
Molecular Weight (g/mol): 163.6
Molecular Formula: C6H10ClNO2
InChi Key: FGNUNVVTHHKDAM-UHFFFAOYSA-N
SMILES: Cl.OC(=O)C1=CCCNC1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.