accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 7209-11-2

CH3OSBr

CAS RN 7209-11-2

IUPAC Name: 1-(4-bromothiophen-2-yl)ethan-1-one
Synonyms: 1-(4-bromothiophen-2-yl)ethanone
Molecular Weight (g/mol): 205.07
Molecular Formula: C6H5BrOS
InChi Key: HXVLWNKFMNRJED-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Br)=CS1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.