accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 77613-65-1

FFBr

CAS RN 77613-65-1

IUPAC Name: 2-(bromomethyl)-1,1-difluorocyclopropane
Synonyms: 2-(bromomethyl)-1,1-difluorocyclopropane
Molecular Weight (g/mol): 170.99
Molecular Formula: C4H5BrF2
InChi Key: TUDJNSKRXIUOAJ-UHFFFAOYNA-N
SMILES: FC1(F)CC1CBr
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.