accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 7785-84-4

NaNaNaOPOOPOOOPOOO

CAS RN 7785-84-4

IUPAC Name: trisodium trioxo-1,3,5,2λ⁵,4λ⁵,6λ⁵-trioxatriphosphinane-2,4,6-tris(olate)
Synonyms: sodium trimetaphosphate
Molecular Weight (g/mol): 305.88
Molecular Formula: Na3O9P3
InChi Key: UGTZMIPZNRIWHX-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].[O-]P1(=O)OP([O-])(=O)OP([O-])(=O)O1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.