accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 79-14-1

HOOHO

CAS RN 79-14-1

IUPAC Name: 2-hydroxyacetic acid
Synonyms: glycolic acid glycocide kyselina hydroxyoctova 2-(14)c-labeled glycolic acid caswell no. 470 1-(14)c-labeled glycolic acid calcium salt glycolic acid monoammonium salt glycolic acid glycollate monopotassium salt glycolic acid
Molecular Weight (g/mol): 76.05
Molecular Formula: C2H4O3
InChi Key: AEMRFAOFKBGASW-UHFFFAOYSA-N
SMILES: OCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (13)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (2)
  • (5)
Boiling Point 
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (8)
  • (4)
  • (1)
  • (1)
  • (13)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.