accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 90-72-2

CH3NH3CNCH3H3COHNCH3H3C

CAS RN 90-72-2

IUPAC Name: 2,4,6-tris[(dimethylamino)methyl]phenol
Synonyms: 2,4,6-tris[(dimethylamino)methyl]phenol
Molecular Weight (g/mol): 265.4
Molecular Formula: C15H27N3O
InChi Key: AHDSRXYHVZECER-UHFFFAOYSA-N
SMILES: CN(C)CC1=CC(CN(C)C)=C(O)C(CN(C)C)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.