accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 952-23-8

NNH2H2NHCl

CAS RN 952-23-8

IUPAC Name: dihydrogen bis(acridine-3,6-diamine) hydrate dichloride
Synonyms: dihydrogen bis(proflavine) hydrate dichloride
Molecular Weight (g/mol): 509.44
Molecular Formula: C26H26Cl2N6O
InChi Key: APOVFGFHTIKDOX-UHFFFAOYSA-N
SMILES: [H+].[H+].O.[Cl-].[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.