accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 98886-44-3

CH3OPOSH3CH3CNSO

CAS RN 98886-44-3

IUPAC Name: ethyl (butan-2-ylsulfanyl)(2-oxo-1,3-thiazolidin-3-yl)phosphinate
Synonyms: fosthiazate nemathorin
Molecular Weight (g/mol): 283.34
Molecular Formula: C9H18NO3PS2
InChi Key: DUFVKSUJRWYZQP-UHFFFAOYNA-N
SMILES: CCOP(=O)(SC(C)CC)N1CCSC1=O
Concentration 
  • (3)
  • (3)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.