accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 99105-77-8

CH3SOOClOOO

CAS RN 99105-77-8

IUPAC Name: 2-(2-chloro-4-methanesulfonylbenzoyl)cyclohexane-1,3-dione
Synonyms: ICIA0051 (sulcotrione)
Molecular Weight (g/mol): 328.76
Molecular Formula: C14H13ClO5S
InChi Key: PQTBTIFWAXVEPB-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(C(=O)C2C(=O)CCCC2=O)C(Cl)=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.