Conversion Tables
Temperature Conversion Tables
Known temperature to be converted in centre column.
Centigrade conversion to left, Fahrenheit to right.
Example
–12.2 10 50
therefore
10°C = 50°F
Temperature Transmittance-Absorbance Units
°CKnown°F %TA.U.%TA.U.
–17,8032 1251.292
–15541 21,69952.284
–12,21050 31,52353.276
–9,41559 41,39854.268
–6,72068 51,30155.260
–3,92577 61,22256.252
–1,13086 71,15557.241
4,440104 81,09758.237
1050122 91,04659.299
15,660140 10160.222
21,170158 11.95961.215
26,780176 12.92162.208
32,290194 13.88663.201
37,8100212 14.85464.194
43,3110230 15.82465.187
48,9120248 16.79666.180
54,4130266 17.77067.174
60140284 18.74568.167
65,6150302 19.72169.161
71,1160320 20.69970.155
76,7170338 21.67871.149
82,2180356 22.65872.143
87,8190374 23.63873.137
93,3200392 24.62074.131
98,9210410 25.60275.125
104,4220428 26.58576.119
110230446 27.56977.114
115,6240464 28.53378.108
121250482 29.53879.102
149300572 30.52380.097
177350662 31.50981.092
204400752 32.49582.086
232450842 33.48283.081
260500932 34.46984.076
2885501022 35.46985.071
3166001112 36.44486.066
3436501202 37.43287.060
3717001292 38.42088.056
3997501382 39.40989.051
4278001472 40.39890.046
4548501562 41.38791.041
4829001652 42.37792.036
5109501742 43.36793.032
53810001832 44.35794.027
58610501922 45.34795.022
59311002012 46.33796.018
62111502102 47.32897.013
64912002192 48.31998.009
67712502282 49.31099.004
70413002372 50.301100.000
73213502462     
76014002552     
78814502642     
81615002732     
84315502822     
87116002912     
89916503002     
92717003092     
95417503182     
98218003272     
101018503362     
103819003452     
106619503542     
109320003632     
Constant Humidity Solutions
Saturated solutions of the salts in contact with excess solute produce the humidities indicated in a closed system.
SaltTemperature °C
1520253035405060
Relative Humidity %
Potassium sulfate9797979696969696
Potassium nitrate9493929189888582
Potassium chloride8786858483828180
Ammonium sulfate8181808080797978
Sodium chloride7676757575757575
Sodium nitrite–66656362625959
Ammonium nitrate6965625955534742
Sodium dichromate56555452515047–
Magnesium nitrate5655535250494643
Potassium carbonate444443434342––
Magnesium chloride3433333332323130
Potassium acetate232322222120––
Lithium chloride1312121212111111
Frequently Used Abbreviations
APHAAmerican Public Health Association
CASChemical Abstracts Service
Cs.case
d, Ddextrorotatory
DOTDepartment of Transportation
Ea.Each
F.W.Formula Weight
I, LLaevorotatory
ìmmicrometre (10-6m)
M.W.Molecular weight
nmnanometre (10-9m)
ppbparts per billion
ppmparts per million
RTroom temperature
TSCAToxic Substances Control Act
USPUnited States Pharmacopeia
Codon dictionary
  Middle base  
5’-OH Terminal baseUCAG3’-OH Terminal base
UPhe Phe Leu LeuSer Ser Ser SerTyr Tyr STOP STOPCys Cys STOP TrpU C A G
CLeu Leu Leu LeuPro Pro Pro ProHis His Gln GlnArg Arg Arg ArgU C A G
AIle Ile Ile MetThr Thr Thr ThrAsn Asn Lys LysSer Ser Arg ArgU C A G
GVal Val Val ValAla Ala Ala AlaAsp Asp Glu GluGly Gly Gly GlyU C A G