Thermo Scientific Chemicals L-Tyrosine, 99+%
L-Tyrosine, CAS # 60-18-4, is an aromatic non-essential amino acid that is considered a fundamental metabolite, involved in protein synthesis and photosynthesis. | CAS: 60-18-4 | C9H11NO3 | 181.19 g/mol
Product Details
8098.69 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Additional Details:
Additional Details: CAS Number: 60-18-4 Weight: 2.50000kg
Quantity 2.5 kg
Packaging Plastic Drum
Disclaimers: RUO – Research Use Only
Product Code. 10702744
Quantity Price
1 8140.0  DKK / 2.50kg
In stock, ready to ship: 4
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Chemical Identifiers
CAS 60-18-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002606
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
Synonym l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l
PubChem CID 6057
ChEBI CHEBI:17895
IUPAC Name (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
Specification
Chemical Name or Material L-Tyrosine
CAS 60-18-4
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.5% max. (1 g, 105°C)
Molecular Formula C9H11NO3
Packaging Plastic Drum
Linear Formula 4-(HO)C6H4CH2CH(NH2)CO2H
MDL Number MFCD00002606
Beilstein 14, 605
Specific Rotation -9.8° to -12.3° (20°C, 589nm) (c=5, 1N HCl)
Merck Index 15, 10020
Sulfated Ash 0.1% max.
Synonym l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l
Synonym l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l
Synonym l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l
Synonym l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l
Synonym l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l
Synonym l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l
Synonym l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l
Synonym l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l
Synonym l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l
Synonym l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
SMILES N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
IUPAC Name (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
Specific Rotation Condition − 11.05 (20.00°C c=5,dilHCL/H2O 50/50)
Molecular Weight (g/mol) 181.19
Quantity 2.5 kg
PubChem CID 6057
ChEBI CHEBI:17895
Formula Weight 181.19
Percent Purity ≥99.0% (on dried substance)
Physical Form Crystalline Powder
General description
• L-Tyrosine is the levorotatory isomer of the aromatic amino acid tyrosine, synthesized in vivo from L-phenylalanine
• It is available as a crystalline, off-white powder, stable at room temperature, and soluble in hydrochloric acid
Applications
• In vivo L-tyrosine is involved in protein synthesis and is a precursor to alkaloids such as morphine, neurotransmitters, epinephrine, p-coumaric acid, thyroxine, pigment melanin and catecholamines.
• Used as a supplement, L-tyrosine has been shown to increase levels of the neurotransmitters, dopamine, adrenaline, and norepinephrine in the brain
• It is considered an essential dietary supplement for phenylketonuria patients who lack the enzyme phenylalanine hydroxylase, and hence cannot convert phenylalanine into tyrosine
• L-tyrosine plays a vital role as an electron-carrier in the process of photosynthesis
• Due to its role in the synthesis of catecholamines, L-tyrosine is also used in research to study its impact on stress
Alternative Products
Documentation
Technical documents