missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Thiocarbonyldiimidazole, tech 90%, Thermo Scientific Chemicals

Product Code. 11479487
Click to view available options
Quantity:
10 g
50 g
250 g

1,1'-Thiocarbonyldiimidazole is used as a reagent for the deoxygenation of alcohols with tributyltin hydride and diols with lithium aluminum hydride. It is also used to deoxygenate carboxylic monosaccharide analogues. Further, it is a useful reagent involved in biochemical synthesis. In addition to this, it is used to prepare 1-[bis-(4-methoxy-phenyl)-methyl]-1H-imidazole by reacting with 4,4'-dimethoxy-benzhydrol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1′-Thiocarbonyldiimidazole is used as a reagent for the deoxygenation of alcohols with tributyltin hydride and diols with lithium aluminum hydride. It is also used to deoxygenate carboxylic monosaccharide analogues. Further, it is a useful reagent involved in biochemical synthesis. In addition to this, it is used to prepare 1-[bis-(4-methoxy-phenyl)-methyl]-1H-imidazole by reacting with 4,4′-dimethoxy-benzhydrol.

Solubility
Soluble in terahydrofuran, toluene and dichloromethane.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6160-65-2
Molecular Formula C7H6N4S
Molecular Weight (g/mol) 178.21
MDL Number MFCD00005289
InChI Key RAFNCPHFRHZCPS-UHFFFAOYSA-N
Synonym 1,1'-thiocarbonyldiimidazole, thiocarbonyldiimidazole, n,n'-thiocarbonyldiimidazole, di 1h-imidazol-1-yl methanethione, 1,1-thiocarbonyldiimidazole, di imidazol-1-yl methanethione, n,n'-thiocarbonyldiimidazole tcdi, 1h-imidazole, 1,1'-carbonothioylbis, n,n-thiocarbonyldiimidazole, 1,1'-carbonothioylbis-1h-imidazole
PubChem CID 80264
IUPAC Name di(imidazol-1-yl)methanethione
SMILES S=C(N1C=CN=C1)N1C=CN=C1

Specifications

Melting Point 98°C to 102°C
Assay Percent Range 0.95
Quantity 50 g
Beilstein 609349
Sensitivity Moisture sensitive
Solubility Information Soluble in terahydrofuran,toluene and dichloromethane.
Formula Weight 178.22
Percent Purity 90%
Grade Technical
Chemical Name or Material 1,1'-Thiocarbonyldiimidazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.