missing translation for 'onlineSavingsMsg'
Learn More
(+)-1,2-Bis[(2S,5S)-2,5-diethyl-1-phospholanyl]benzene, 97+% Product Code.: 15476156

(+)-1,2-Bis[(2S,5S)-2,5-diethyl-1-phospholanyl]benzene, 97+%

Product Code. 15476156
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15476156

Brand: Thermo Scientific Alfa Aesar H60981.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 136779-28-7 | C22H36P2 | 362.478 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136779-28-7
Molecular Formula C22H36P2
Molecular Weight (g/mol) 362.478
MDL Number MFCD00142321
InChI Key GVVCHDNSTMEUCS-MUGJNUQGSA-N
Synonym ethyl-duphos, s,s, unii-fri8026kma, s,s-et-duphos, fri8026kma, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene, +-duphos, 2s,5s-1-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane, s,s-ethyl-duphosShow MoreShow Less
PubChem CID 5702640
IUPAC Name (2S,5S)-1-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane
SMILES CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC

Specifications

Density 1.01
Refractive Index 1.581
Quantity 1 g
Beilstein 4814174
Sensitivity Air Sensitive
Optical Rotation +265° (c=1 in Hexane)
Formula Weight 362.47
Percent Purity ≥97%
Chemical Name or Material (+)-1,2-Bis[(2S,5S)-2,5-diethyl-1-phospholanyl]benzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP25K030 Certificate TypeCertificate of Analysis Date05/04/2024 Catalog NumberH60981.03,H60981.MD
Lot NumberR15J010 Certificate TypeCertificate of Analysis Date22/05/2023 Catalog NumberH60981.03
Lot NumberN09H036 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60981.03,H60981.MD
Lot NumberK27Y077 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60981.03,H60981.MD
Lot NumberZ18D051 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60981.03,H60981.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(+)-1,2-Bis[(2S,5S)-2,5-diethyl-1-phospholanyl]benzene, 97+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.