Learn More
1,2-Bis(diphenylphosphino)benzene, 98% Product Code.: 11454567
1,2-Bis(diphenylphosphino)benzene, 98%
Product Code. 11454567
1 g
Click to view available options
Quantity:
0.25 g
1 g
Product Code. 11454567
Brand: Thermo Scientific Alfa Aesar B21106.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 13991-08-7 | C30H24P2 | 446.47 g/mol
It acts as a ligand for Baeyer-Villiger Oxidation, cycloadditions and reductions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It acts as a ligand for Baeyer-Villiger Oxidation, cycloadditions and reductions.
Solubility
Insoluble in water.
Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13991-08-7
Molecular Formula C30H24P2
Molecular Weight (g/mol) 446.47
MDL Number MFCD00014081
InChI Key NFRYVRNCDXULEX-UHFFFAOYSA-N
Synonym 1,2-bis diphenylphosphino benzene, dppbz, dppbe, dppben, dppbenz, o-bis diphenylphosphino benzene, 1,2-bis dimethylphosphino benzene, o-phenylenebis diphenylphosphine, phosphine, phenylenebis diphenyl, 1,2-bis diphenylphosphanyl benzene
PubChem CID 498379
IUPAC Name (2-diphenylphosphanylphenyl)-diphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Melting Point 185°C to 189°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 446.47
Percent Purity 98%
Chemical Name or Material 1,2-Bis(diphenylphosphino)benzene
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459232 Certificate TypeCertificate of Analysis Date15/11/2024 Catalog NumberB21106.03,B21106.01
Lot NumberA0459738 Certificate TypeCertificate of Analysis Date13/09/2024 Catalog NumberB21106.03,B21106.01
Lot Number10224764 Certificate TypeCertificate of Analysis Date13/07/2023 Catalog NumberB21106.03,B21106,B21106.01
Lot Number10148613 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21106.03,B21106.01
Lot NumberG19G04 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21106.03,B21106.01
RUO – Research Use Only
Product Content Correction
Product Title
1,2-Bis(diphenylphosphino)benzene, 98% >