missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diaminocyclohexane, mixture of isomers, 99%, Thermo Scientific Chemicals

Product Code. 11470518
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,2-Diaminocyclohexane was used in the synthesis of chiral ruthenium(IV)-oxo complexes & bicyclic pyrazine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diaminocyclohexane was used in the synthesis of chiral ruthenium(IV)-oxo complexes & bicyclic pyrazine derivatives.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, air, carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-83-7
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00001491
InChI Key SSJXIUAHEKJCMH-UHFFFAOYSA-N
Synonym 1,2-cyclohexanediamine, 1,2-diaminocyclohexane, 1,2-cylohexanediamine, cyclohex-1,2-ylenediamine, 1r,2r---1,2-diamino cyclohexane, dsstox_cid_7301, dsstox_rid_78396, dsstox_gsid_27301, +/--trans-1,2-diaminocyclohexane, cis-1,2-cyclohexandiamine
PubChem CID 4610
IUPAC Name cyclohexane-1,2-diamine
SMILES C1CCC(C(C1)N)N

Specifications

Density 0.94
pH 12
Boiling Point 191°C
Flash Point 75°C (167°F)
Odor Amine-like
Refractive Index 1.487
Quantity 1000 g
UN Number UN2735
Beilstein 506142
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material 1,2-Diaminocyclohexane, mixture of isomers
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2-Diaminocyclohexane, mixture of isomers
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-Cyclohexanediamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'