missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(Dimethylamino)propyl]-3-ethylurea, 97%, Thermo Scientific Chemicals

407.16 DKK - 4468.96 DKK

Chemical Identifiers

CAS 32897-26-0
Molecular Formula C8H19N3O
Molecular Weight (g/mol) 173.26
MDL Number MFCD00027752
InChI Key NGJUYARYEXGDNN-UHFFFAOYSA-N
Synonym 1-3-dimethylamino propyl-3-ethylurea, 1-ethyl-3 3-dimethylamino urea, 1-ethyl-3-3-dimethylaminopropyl urea, urea, n-3-dimethylamino propyl-n'-ethyl, 3-3-dimethylamino propyl-1-ethylurea, 1-3-dimethylamino propyl-3-ethyl isourea, 1-3-dimethylaminopropyl-3-ethylurea, 3-ethyl amino hydroxy methylene amino-n,n-dimethylpropan-1-amine
PubChem CID 4196313
IUPAC Name 1-[3-(dimethylamino)propyl]-3-ethylurea
SMILES CCNC(=O)NCCCN(C)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15410769
View Documents
Thermo Scientific Alfa Aesar
H63845.MD
250 mg
407.16 DKK
250mg
Please to purchase this item. Need a web account? Register with us today!
15490759
View Documents
Thermo Scientific Alfa Aesar
H63845.03
1 g
1222.22 DKK
1g
Please to purchase this item. Need a web account? Register with us today!
15400769
View Documents
Thermo Scientific Alfa Aesar
H63845.06
5 g
4468.96 DKK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-[3-(Dimethylamino)propyl]-3-ethylurea is used in the preparation of cabergoline, which is a dopamine receptor used to treat Parkinsons syndrom.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-[3-(Dimethylamino)propyl]-3-ethylurea is used in the preparation of cabergoline, which is a dopamine receptor used to treat Parkinsons syndrom.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

32897-26-0
173.26
NGJUYARYEXGDNN-UHFFFAOYSA-N
4196313
CCNC(=O)NCCCN(C)C
C8H19N3O
MFCD00027752
1-3-dimethylamino propyl-3-ethylurea, 1-ethyl-3 3-dimethylamino urea, 1-ethyl-3-3-dimethylaminopropyl urea, urea, n-3-dimethylamino propyl-n'-ethyl, 3-3-dimethylamino propyl-1-ethylurea, 1-3-dimethylamino propyl-3-ethyl isourea, 1-3-dimethylaminopropyl-3-ethylurea, 3-ethyl amino hydroxy methylene amino-n,n-dimethylpropan-1-amine
1-[3-(dimethylamino)propyl]-3-ethylurea

Specifications

32897-26-0
CH3CH2NHCONH(CH2)3N(CH3)2
4961600
Soluble in water.
CCNC(=O)NCCCN(C)C
173.26
173.26
1-[3-(Dimethylamino)propyl]-3-ethylurea
C8H19N3O
MFCD00027752
1-3-dimethylamino propyl-3-ethylurea, 1-ethyl-3 3-dimethylamino urea, 1-ethyl-3-3-dimethylaminopropyl urea, urea, n-3-dimethylamino propyl-n'-ethyl, 3-3-dimethylamino propyl-1-ethylurea, 1-3-dimethylamino propyl-3-ethyl isourea, 1-3-dimethylaminopropyl-3-ethylurea, 3-ethyl amino hydroxy methylene amino-n,n-dimethylpropan-1-amine
NGJUYARYEXGDNN-UHFFFAOYSA-N
1-[3-(dimethylamino)propyl]-3-ethylurea
4196313
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • 1-[3-(Dimethylamino)propyl]-3-ethylurea

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 1-[3-(Dimethylamino)propyl]-3-ethylurea

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.