missing translation for 'onlineSavingsMsg'
Learn More
1,3-thiazol-2-ylmethylamine, 97%, Thermo Scientific™ Product Code.: 10736105

1,3-thiazol-2-ylmethylamine, 97%, Thermo Scientific™

Product Code. 10736105
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10736105

Brand: Thermo Scientific Maybridge CC09513DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 850852-85-6
Molecular Formula C4H7ClN2S
Molecular Weight (g/mol) 150.624
MDL Number MFCD02854204
InChI Key AAJCCSYSPIKAJY-UHFFFAOYSA-N
Synonym thiazol-2-ylmethanamine hydrochloride, 2-aminomethylthiazole hcl, 1,3-thiazol-2-ylmethanamine hydrochloride, 2-aminomethylthiazole hydrochloride, 2-amino methylthiazole hydrochloride, c-thiazol-2-yl-methylamine hydrochloride, 2-thiazolemethanamine hydrochloride, 1,3-thiazol-2-ylmethylamine hydrochloride, 2-aminomethyl thiazole hydrochloride, 1,3-thiazol-2-ylmethylamine, chloride
PubChem CID 42614216
IUPAC Name 1,3-thiazol-2-ylmethanamine;hydrochloride
SMILES C1=CSC(=N1)CN.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 150.63
Percent Purity 97%
Chemical Name or Material 1, 3-Thiazol-2-ylmethylamine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-thiazol-2-ylmethylamine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.