missing translation for 'onlineSavingsMsg'
Learn More
1,4-Bis(2-hydroxyethyl)piperazine, 98% Product Code.: 15491188

1,4-Bis(2-hydroxyethyl)piperazine, 98%

Product Code. 15491188
25 g, 25g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15491188

Brand: Thermo Scientific Alfa Aesar H55063.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-96-3 | C8H18N2O2 | 174.244 g/mol

It is applied for desulfurization, decarburization extraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied for desulfurization, decarburization extraction.

Solubility
Soluble in water.

Notes
Store at 4°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-96-3
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.244
MDL Number MFCD00006157
InChI Key VARKIGWTYBUWNT-UHFFFAOYSA-N
Synonym 1,4-piperazinediethanol, 2,2'-piperazine-1,4-diyl diethanol, 1,4-bis 2-hydroxyethyl piperazine, n,n'-bis 2-hydroxyethyl piperazine, 1,4-di 2-hydroxyethyl piperazine, n,n'-dihydroxyethylpiperazine, n,n'-di 2-hydroxyethyl piperazine, unii-eq349pkx8y, piperazine, n,n'-bis 2-hydroxyethyl, 1,4-bis beta-hydroxyethyl piperazine
PubChem CID 67151
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanol
SMILES C1CN(CCN1CCO)CCO

Specifications

Melting Point 133°C to 136°C
Boiling Point 215°C to 220°C (50 mmHg)
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 174.24
Percent Purity 99%
Chemical Name or Material 1,4-Bis(2-hydroxyethyl)piperazine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberU30K025 Certificate TypeCertificate of Analysis Date16/09/2024 Product CodeH55063.14,H55063.06
Lot NumberQ02K008 Certificate TypeCertificate of Analysis Date10/04/2024 Product CodeH55063.14,H55063.06
Lot NumberP09H029 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH55063.14,H55063.06
Lot NumberS13E004 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH55063.14,H55063.06
Lot NumberW10E037 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH55063.14,H55063.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4-Bis(2-hydroxyethyl)piperazine, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.