missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Iodo-2-pyridyl)piperazine, 95%, Thermo Scientific™

Product Code. 11342349
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 219635-89-9
Molecular Formula C9H12IN3
Molecular Weight (g/mol) 289.12
MDL Number MFCD04039867
InChI Key MKKJIJFTSAYHBW-UHFFFAOYSA-N
Synonym 1-5-iodopyridin-2-yl piperazine, 1-5-iodo-pyridin-2-yl-piperazine, 1-5-iodo-2-pyridyl piperazine, 1-5-iodo-2-pyridinyl piperazine, 4-5-iodopyridin-2-yl piperazine, piperazine, 1-5-iodo-2-pyridinyl, acmc-20ap9y, 1-5-iodopyridin-2-yl-piperazine, 1-5-iodanylpyridin-2-yl piperazine, piperazine,1-5-iodo-2-pyridinyl
PubChem CID 17750162
IUPAC Name 1-(5-iodopyridin-2-yl)piperazine
SMILES IC1=CC=C(N=C1)N1CCNCC1

Specifications

Melting Point ∽135°C (decomposition)
Quantity 1 g
Sensitivity Air and light sensitive
Formula Weight 289.12
Percent Purity 95%
Chemical Name or Material 1-(5-Iodo-2-pyridyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'