missing translation for 'onlineSavingsMsg'
Learn More
Alfa Aesar™ 1-Boc-6-amino-1H-indazole, 97% Product Code.: 15444677

Alfa Aesar™ 1-Boc-6-amino-1H-indazole, 97%

Product Code. 15444677
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15444677

Brand: Alfa Aesar™ H32010.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
Used in the synthesis of chiral molecules.

Solubility
Slightlysoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 219503-81-8
Molecular Formula C12H15N3O2
Molecular Weight (g/mol) 233.27
MDL Number MFCD04114657
InChI Key ZUBZKAXFZUIBQY-UHFFFAOYSA-N
Synonym 1-boc-6-amino-1h-indazole, tert-butyl 6-amino-1h-indazole-1-carboxylate, 1-boc-6-amino-indazole, 1-boc-6-aminoindazole, 6-amino-1h-indazole-1-carboxylic acid, 1,1 dimethyl ethyl ester, 1h-indazole-1-carboxylic acid, 6-amino-, 1,1-dimethylethyl ester, acmc-1cff2, ksc494o9t, 1-boc-1h-indazol-6-amine, 6-amino-1h-indazole, n1-boc protected
PubChem CID 12018607
IUPAC Name tert-butyl 6-aminoindazole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1N=CC2=CC=C(N)C=C12

Specifications

Melting Point 171°C to 172°C
Quantity 1 g
Solubility Information Slightlysoluble in water.
Formula Weight 233.27
Percent Purity 97%
Chemical Name or Material 1-Boc-6-amino-1H-indazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Alfa Aesar™ 1-Boc-6-amino-1H-indazole, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.