missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-iodoethane, 96%, stab. with copper, Thermo Scientific Chemicals

312.68 DKK - 4191.53 DKK

Chemical Identifiers

CAS 624-70-4
Molecular Formula C2H4ClI
Molecular Weight (g/mol) 190.41
MDL Number MFCD00041453
InChI Key JTWWWQGSFTWWDL-UHFFFAOYSA-N
Synonym 1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb
PubChem CID 136438
IUPAC Name 1-chloro-2-iodoethane
SMILES ClCCI
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11302976
View Documents
Thermo Scientific Alfa Aesar
L10742.03
1 g
312.68 DKK
1g
Please to purchase this item. Need a web account? Register with us today!
11312976
View Documents
Thermo Scientific Alfa Aesar
L10742.06
5 g
1072.25 DKK
5g
Please to purchase this item. Need a web account? Register with us today!
11322976
View Documents
Thermo Scientific Alfa Aesar
L10742.14
25 g
4191.53 DKK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.

Solubility
Soluble inorganic solvents. Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

624-70-4
190.41
JTWWWQGSFTWWDL-UHFFFAOYSA-N
136438
ClCCI
C2H4ClI
MFCD00041453
1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb
1-chloro-2-iodoethane

Specifications

624-70-4
61°C to 62°C (30 mmHg)
C2H4ClI
MFCD00041453
Light sensitive
Soluble inorganic solvents. Insoluble in water.
ClCCI
190.41
190.41
1-Chloro-2-iodoethane
2.088
>100°C (212°F)
1.566
1730865
1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb
JTWWWQGSFTWWDL-UHFFFAOYSA-N
1-chloro-2-iodoethane
136438
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 677-827-4

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only