missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-nitrobenzene, 99%, Thermo Scientific Chemicals

Product Code. 11418376
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

1-Chloro-2-nitrobenzene was used in the synthesis of 1-hydroxybenzotriazole derivatives. It is important as a precursor to other compounds due to the two reactive sites present on the molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-nitrobenzene was used in the synthesis of 1-hydroxybenzotriazole derivatives. It is important as a precursor to other compounds due to the two reactive sites present on the molecule.

Solubility
Soluble in water 0.59 g/L.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-73-3
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.55
MDL Number MFCD00007061
InChI Key BFCFYVKQTRLZHA-UHFFFAOYSA-N
Synonym 2-chloronitrobenzene, o-chloronitrobenzene, 2-nitrochlorobenzene, chloronitrobenzene, o-nitrochlorobenzene, benzene, 1-chloro-2-nitro, 2-chloro-1-nitrobenzene, oncb, benzene, chloronitro, nitrochlorobenzene
PubChem CID 6945
ChEBI CHEBI:34270
IUPAC Name 1-chloro-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC=C1Cl

Specifications

Melting Point 31°C to 34°C
Density 1.348
Boiling Point 245°C to 246°C
Flash Point 126°C (258°F)
Quantity 1000 g
UN Number UN1578
Beilstein 509273
Merck Index 14,2151
Solubility Information Soluble in water 0.59g/L.
Formula Weight 157.56
Percent Purity 99%
Chemical Name or Material 1-Chloro-2-nitrobenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.