Learn More
1-Chloro-3-methyl-2-butene, 95%, stab. with potassium carbonate, Thermo Scientific Chemicals
698.84 DKK - 2234.49 DKK
Chemical Identifiers
CAS | 503-60-6 |
---|---|
Molecular Formula | C5H9Cl |
Molecular Weight (g/mol) | 104.577 |
MDL Number | MFCD00000982 |
InChI Key | JKXQKGNGJVZKFA-UHFFFAOYSA-N |
Synonym | 1-chloro-3-methyl-2-butene, prenyl chloride, 2-butene, 1-chloro-3-methyl, 3,3-dimethylallyl chloride, 3-methyl-2-butenyl chloride, 3-methylcrotyl chloride, unii-kmt7v9llbr, gamma,gamma-dimethylallyl chloride, kmt7v9llbr, .gamma.,.gamma.-dimethylallyl chloride |
PubChem CID | 10429 |
IUPAC Name | 1-chloro-3-methylbut-2-ene |
SMILES | CC(=CCCl)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11447867
|
Thermo Scientific Alfa Aesar
A16545.09 |
10 g | N/A | N/A | N/A | |||||
11457867
|
Thermo Scientific Alfa Aesar
A16545.18 |
50 g |
2234.49 DKK
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
1-Chloro-3-methyl-2-butene is used in the synthesis of geraniol. It is also used as an alkylating agent and as a reagent in hyperforin. Further, it is used as an antibiotic to inhibit growth of tumor cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Chloro-3-methyl-2-butene is used in the synthesis of geraniol. It is also used as an alkylating agent and as a reagent in hyperforin. Further, it is used as an antibiotic to inhibit growth of tumor cells.
Solubility
Miscible with chloroform, acetone, diethyl ether and alcohol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids. Store in a cool place.
Chemical Identifiers
503-60-6 | |
104.577 | |
JKXQKGNGJVZKFA-UHFFFAOYSA-N | |
10429 | |
CC(=CCCl)C |
C5H9Cl | |
MFCD00000982 | |
1-chloro-3-methyl-2-butene, prenyl chloride, 2-butene, 1-chloro-3-methyl, 3,3-dimethylallyl chloride, 3-methyl-2-butenyl chloride, 3-methylcrotyl chloride, unii-kmt7v9llbr, gamma,gamma-dimethylallyl chloride, kmt7v9llbr, .gamma.,.gamma.-dimethylallyl chloride | |
1-chloro-3-methylbut-2-ene |
Specifications
503-60-6 | |
107°C to 109°C | |
C5H9Cl | |
1.449 | |
UN1993 | |
Miscible with chloroform,acetone,diethyl ether and alcohol. Immiscible with water. | |
CC(=CCCl)C | |
104.577 | |
104.57 | |
1-Chloro-3-methyl-2-butene, stabilized with potassium carbonate |
0.928 | |
13°C (56°F) | |
(CH3)2C=CHCH2Cl | |
MFCD00000982 | |
1-chloro-3-methyl-2-butene, prenyl chloride, 2-butene, 1-chloro-3-methyl, 3,3-dimethylallyl chloride, 3-methyl-2-butenyl chloride, 3-methylcrotyl chloride, unii-kmt7v9llbr, gamma,gamma-dimethylallyl chloride, kmt7v9llbr, .gamma.,.gamma.-dimethylallyl chloride | |
JKXQKGNGJVZKFA-UHFFFAOYSA-N | |
1-chloro-3-methylbut-2-ene | |
10429 | |
95% |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 207-972-7
missing translation for 'rtecsNumber' : EM4298500
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.