missing translation for 'onlineSavingsMsg'
Learn More

1-Chloropropane, 99%, Thermo Scientific Chemicals

290.18 DKK - 1282.20 DKK

Chemical Identifiers

CAS 540-54-5
Molecular Formula C3H7Cl
Molecular Weight (g/mol) 78.54
MDL Number MFCD00000995
InChI Key SNMVRZFUUCLYTO-UHFFFAOYSA-N
Synonym propyl chloride, propane, 1-chloro, n-propyl chloride, chloropropane, unii-tuv7462nwk, 1-chloro-propane, tuv7462nwk, n-propylchloride, propane, chloro, 1-chloropropyl
PubChem CID 10899
IUPAC Name 1-chloropropane
SMILES CCCCl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10244470
View Documents
Thermo Scientific Acros
109950050
5 mL Glass bottle
290.18 DKK
5mL
Please to purchase this item. Need a web account? Register with us today!
10794211
View Documents Promotion Details
Thermo Scientific Acros
109951000
100 mL Glass bottle
402.65 DKK
100mL
Please to purchase this item. Need a web account? Register with us today!
10259223
View Documents
Thermo Scientific Acros
109955000
500 mL Glass bottle
1282.20 DKK
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

540-54-5
78.54
SNMVRZFUUCLYTO-UHFFFAOYSA-N
10899
CCCCl
C3H7Cl
MFCD00000995
propyl chloride, propane, 1-chloro, n-propyl chloride, chloropropane, unii-tuv7462nwk, 1-chloro-propane, tuv7462nwk, n-propylchloride, propane, chloro, 1-chloropropyl
1-chloropropane

Specifications

540-54-5
100.0
Colorless
46°C to 47°C
Authentic
C3H7Cl
1.3870 to 1.389
01, 104
0.892
Solubility in water: 2.7g/L (20°C). Other solubilities: soluble in alcohol,ether and benzene
CCCCl
78.54
78.54
Liquid
98.5
-123°C
0.8920g/mL
-18°C
98.5% min. (GC)
CH3CH2CH2Cl
MFCD00000995
15, 7960
propyl chloride, propane, 1-chloro, n-propyl chloride, chloropropane, unii-tuv7462nwk, 1-chloro-propane, tuv7462nwk, n-propylchloride, propane, chloro, 1-chloropropyl
SNMVRZFUUCLYTO-UHFFFAOYSA-N
1-chloropropane
10899
99%
1-chloropropane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 208-749-7

Product Identifier
  • 1-Chloropropane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only