missing translation for 'onlineSavingsMsg'
Learn More

1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt, 95%, Thermo Scientific™

197.48 DKK - 2492.40 DKK

Chemical Identifiers

CAS 136997-71-2
Molecular Formula C10H14N2O4S
Molecular Weight (g/mol) 258.29
MDL Number MFCD00185917
InChI Key SJHHXGGXZKPIDE-UHFFFAOYSA-N
Synonym 1-n,n-dimethylcarbamoyl-4-2-sulfoethyl pyridinium hydroxide inner salt, 1-dimethylcarbamoyl-4-2-sulfoethyl pyridinium inner salt, 2-1-dimethylcarbamoyl pyridin-1-ium-4-yl ethanesulfonate, 1-dimethylcarbamoyl-4-2-sulfoethyl pyridinium hydroxide inner salt, 1-dimethylcarbamoyl-4-2-sulfonatoethyl pyridin-1-ium, 2-1-n,n-dimethylcarbamoyl-4-pyridyl ethanesulfonic acid, 1-dimethylcarbamoyl-4-2-sulfoethyl pyridinium betaine, 1-dimethylcarbamoyl-4-2-sulfonatoethyl pyridinium, 2-1-dimethylcarbamoyl-4-pyridinio ethylsulfonate
PubChem CID 3853422
SMILES CN(C)C(=O)[N+]1=CC=C(CCS([O-])(=O)=O)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11479637
SDS View Documents
Thermo Scientific Alfa Aesar
B22124.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11489637
SDS View Documents
Thermo Scientific Alfa Aesar
B22124.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11499637
SDS View Documents
Thermo Scientific Alfa Aesar
B22124.30
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

missing translation for 'einecsNumber' : 418-440-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt, 95%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.