Learn More
(+/-)-1-Ferrocenylethanol, 99%, Thermo Scientific Chemicals Product Code.: 11446860
(+/-)-1-Ferrocenylethanol, 99%, Thermo Scientific Chemicals
Product Code. 11446860
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11446860
Brand: Thermo Scientific Alfa Aesar L19681.06
Request Bulk or Custom Format
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
(±)-1-Ferrocenylethanol is a ligand in titanium-catalyzed intermolecular hydroamination of terminal alkynes (Markovnikov vs. anti-Markovnikov addition). Used as a reactant for single-electron transfer living radical polymerization reactions, preparation of ferrocene-modified thiopyrimidines as anticancer agents, preparation of rersible addition-fragmentation chain transfer (RAFT) agent for RAFT polymerization of styrene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(+/-)-1-Ferrocenylethanol is a ligand in titanium-catalyzed intermolecular hydroamination of terminal alkynes (Markovnikov vs. anti-Markovnikov addition). Used as a reactant for single-electron transfer living radical polymerization reactions, preparation of ferrocene-modified thiopyrimidines as anticancer agents, preparation of rersible addition-fragmentation chain transfer (RAFT) agent for RAFT polymerization of styrene.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1277-49-2
Molecular Formula C12H14FeO
Molecular Weight (g/mol) 230.09
MDL Number MFCD00013752
InChI Key KGIBJAFJTDOHBV-LURJTMIESA-N
Synonym 1-Hydroxyethylferrocene; alpha-Methylferrocenemethanol
IUPAC Name (±)-1-Ferrocenylethanol
SMILES [Fe].c1cccc1.CC(O)c1cccc1
Specifications
Melting Point 77°C to 79°C
Flash Point >100°C (212°F)
Quantity 5 g
Formula Weight 230.09
Percent Purity 99%
Chemical Name or Material (+/-)-1-Ferrocenylethanol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10145344 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL19681.06,L19681.03
Lot Number10178847 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL19681.06,L19681.03
Lot Number10145344 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL19681.06,L19681.03
Lot Number10178847 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL19681.06,L19681.03
RUO – Research Use Only
Product Content Correction
Product Title
(+/-)-1-Ferrocenylethanol, 99%, Thermo Scientific Chemicals >