Learn More
1-Hexyn-3-ol, 97%, Thermo Scientific™ Product Code.: 11302487
1-Hexyn-3-ol, 97%, Thermo Scientific™
Product Code. 11302487
5 g
Click to view available options
Quantity:
5 g
25 g
Product Code. 11302487
Brand: Thermo Scientific Alfa Aesar L11549.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemische identificatiekenmerken
CAS 105-31-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00014408
InChI Key LTFTWJYRQNTCHI-UHFFFAOYSA-N
Synonym 1-hexyn-3-ol, propylethinylcarbinol, 1-hexyne-3-ol, 3-hydroxy-1-hexyne, 1-propylpropargyl alcohol, 1-hexine-3-ol, acmc-2098fm, dsstox_cid_30088, dsstox_gsid_51536, 4-01-00-02234 beilstein handbook referenceShow MoreShow Less
PubChem CID 7746
IUPAC Name hex-1-yn-3-ol
SMILES CCCC(C#C)O
Specificatie
Density 0.872
Boiling Point 142°C to 143°C
Flash Point 47°C (116°F)
Odor Recognizable
Refractive Index 1.435
Quantity 5 g
UN Number UN2929
Beilstein 1739419
Formula Weight 98.15
Percent Purity 97%
Chemical Name or Material 1-Hexyn-3-ol
Meer weergeven Minder weergeven
Product Content Correction
Product Title
1-Hexyn-3-ol, 97%, Thermo Scientific™ >