Learn More
1-Iodo-3-methylbutane, 97%, stab. with copper
CAS: 541-28-6 | C5H11I | 198.047 g/mol
342.00 DKK - 1095.00 DKK
Chemical Identifiers
CAS | 541-28-6 |
---|---|
Molecular Formula | C5H11I |
Molecular Weight (g/mol) | 198.047 |
MDL Number | MFCD00039414 |
InChI Key | BUZZUHJODKQYTF-UHFFFAOYSA-N |
Synonym | isoamyl iodide, isopentyl iodide, butane, 1-iodo-3-methyl, 3-methylbutyliodide, 1-iodo-3-methyl-butane, 1-jod-3-methylbutan, iso-amyliodide, unii-e1e5du187l, 1-jod-3-methylbutan czech, 1-iodo-3-methyl butaneShow More |
PubChem CID | 10924 |
IUPAC Name | 1-iodo-3-methylbutane |
SMILES | CC(C)CCI |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11361847
|
Thermo Scientific Alfa Aesar
L05194.09 |
10 g |
342.00 DKK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371847
|
Thermo Scientific Alfa Aesar
L05194.18 |
50 g |
1095.00 DKK
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Iodo-3-methylbutane, 97%, stab. with copper can be used for organic synthesis or used as solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Iodo-3-methylbutane, 97%, stab. with copper can be used for organic synthesis or used as solvents.
Solubility
Insoluble in water. Soluble in most common organic solvents.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Chemical Identifiers
541-28-6 | |
198.047 | |
BUZZUHJODKQYTF-UHFFFAOYSA-N | |
10924 | |
CC(C)CCI |
C5H11I | |
MFCD00039414 | |
isoamyl iodide, isopentyl iodide, butane, 1-iodo-3-methyl, 3-methylbutyliodide, 1-iodo-3-methyl-butane, 1-jod-3-methylbutan, iso-amyliodide, unii-e1e5du187l, 1-jod-3-methylbutan czech, 1-iodo-3-methyl butaneShow More | |
1-iodo-3-methylbutane |
Specifications
541-28-6 | |
146°C to 148°C | |
C5H11I | |
MFCD00039414 | |
1730990 | |
14,5120 | |
Insoluble in water. Soluble in most common organic solvents. | |
CC(C)CCI | |
198.047 | |
198.05 | |
1-Iodo-3-methylbutane, stabilized with copper |
1.515 | |
40°C (104°F) | |
1.492 | |
UN1993 | |
Light sensitive | |
isoamyl iodide, isopentyl iodide, butane, 1-iodo-3-methyl, 3-methylbutyliodide, 1-iodo-3-methyl-butane, 1-jod-3-methylbutan, iso-amyliodide, unii-e1e5du187l, 1-jod-3-methylbutan czech, 1-iodo-3-methyl butane | |
BUZZUHJODKQYTF-UHFFFAOYSA-N | |
1-iodo-3-methylbutane | |
10924 | |
97% |
Safety and Handling
GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H226-H302
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 208-773-8
missing translation for 'rtecsNumber' : EK4412000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.