Learn More
Fisher Chemical 1 LT METHANOL HPLC 1L
Brand: Fisher Chemical M/4056/15X
1 LT METHANOL HPLC 1L
Description

Safety and Handling
- Methanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P240-Ground/bond container and receiving equipment.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2536521 | Certificate of Analysis | 01/05/2025 | M/4056/25SS,M/4056/99GSK,M/4056/10RSS,M/4056/30RSS,M/4056/27SS,M/4056/34X,M/4056/25RSS,M/4056/99W,M/4056/15Z,M/4056/27J,M/4056/17,M/4056/15X |
2530007 | Certificate of Analysis | 23/04/2025 | M/4056/25SS,M/4056/99GSK,M/4056/10RSS,M/4056/30RSS,M/4056/27SS,M/4056/34X,M/4056/25RSS,M/4056/99W,M/4056/15Z,M/4056/27J,M/4056/17,M/4056/15X |
2533047 | Certificate of Analysis | 03/04/2025 | M/4056/25SS,M/4056/99GSK,M/4056/10RSS,M/4056/30RSS,M/4056/27SS,M/4056/34X,M/4056/25RSS,M/4056/99W,M/4056/15Z,M/4056/27J,M/4056/17,M/4056/15X |
2532263 | Certificate of Analysis | 27/03/2025 | M/4056/25SS,M/4056/99GSK,M/4056/10RSS,M/4056/30RSS,M/4056/27SS,M/4056/34X,M/4056/25RSS,M/4056/99W,M/4056/15Z,M/4056/27J,M/4056/17,M/4056/15X |
2532459 | Certificate of Analysis | 27/03/2025 | M/4056/25SS,M/4056/99GSK,M/4056/10RSS,M/4056/30RSS,M/4056/27SS,M/4056/34X,M/4056/25RSS,M/4056/99W,M/4056/15Z,M/4056/27J,M/4056/17,M/4056/15X |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.