Learn More
SP Bel-Art™ 10 L Carboy Without Spigot
Shop All SP Scienceware Products
Click to view available options
:
10 L Carboy Without Spigot
Description
- 21 CFR 177.1520 compliant for direct contact with foods and beverages
- USP Class VI certified for demands of biotechnology and pharmaceutical industries
- For storing and dispensing large batches of culture media, distilled water and other solutions
- Excellent chemical resistance
- Large handles for easy transport
- Steam autoclavable at 121 °C
- 83 mm screw cap includes built-in thermo-plastic elastomer washer for positive seal
- Molded-in bung accepts spigot
- Located low for more complete drainage
Specifications
Specifications
Autoclavable | Autoclavable |
Capacity (English) | 2.6 gal. |
Capacity (Metric) | 10 L |
Certifications/Compliance | 21 CFR 177.1520 compliant, USP Class VI certified |
Closure Size | 83 mm |
Closure Type | Screw Cap |
For Use With (Application) | For storing and dispensing large batches of culture media, distilled water and other solutions |
Includes | Screw cap with thermo-plastic elastomer washer |
Material | Polypropylene |
Product Type | Carboy |
Product Content Correction
Product Title
SP Bel-Art™ 10 L Carboy Without Spigot >
Spot an opportunity for improvement?Share a Content Correction