Learn More
Fisher Chemical Potassium HYDROXIDE 86 % FOR ANALYse 10kg
Brand: Fisher Chemical P/5640/68
1895.08 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Potassium HYDROXIDE 86 % FOR ANALYse 10kg
Description
Safety and Handling
- Potassium hydroxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2543289 | Certificate of Analysis | 04/07/2025 | P/5640/60,P/5640/70,P/5640/68,P/5640/53,P/5640/65 |
2543287 | Certificate of Analysis | 04/07/2025 | P/5640/60,P/5640/70,P/5640/68,P/5640/53,P/5640/65 |
2543286 | Certificate of Analysis | 04/07/2025 | P/5640/60,P/5640/70,P/5640/68,P/5640/53,P/5640/65 |
2530121 | Certificate of Analysis | 24/05/2025 | P/5640/60,P/5640/70,P/5640/68,P/5640/53,P/5640/65 |