missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human NMDAR1 Protein
Recombinant Protein
Brand: Novus Biologicals™ H00002902-G01-10ug
This item is not returnable.
View return policy
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.Specifications
Immunoaffinity Purification | |
2902 | |
10 μg | |
glutamate [NMDA] receptor subunit zeta-1, glutamate receptor, ionotropic, N-methyl D-aspartate 1, NMDA1, NMDAR1glutamate [NMDA] receptor subunit zeta 1, NMD-R1, N-methyl-D-aspartate receptor channel, subunit zeta-1, N-methyl-D-aspartate receptor subunit NR1, NR1 | |
Neuronal Cell Markers, Neuroscience, Neurotransmission | |
NMDAR1 |
25 mM Tris-HCl pH8.0 in 2% glycerol. | |
M.W. (Theoretical): 103.2 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
GRIN1 | |
>80% by SDS-PAGE and Coomassie blue staining |