Learn More
Extech RH300-CAL Humidity Calibration Standards, 33% and 75%
Brand:  Cole Parmer 37803-05
Extech RH300-CAL Humidity Calibration Standards, 33% and 75%
Product Code. 15707988
  • 765.00 DKK / Each
Only null left
Estimated Shipment: 19-09-2025
Product Title
Extech RH300-CAL Humidity Calibration Standards, 33% and 75% >