Learn More
Testo™ 176 T3 Four-Channel Temperature Data Logger
Brand: Testo™ 0572 1763
This item is not returnable.
View return policy
Specifications
Temperature Datalogger | |
8 years at 15 min. meas. rate, +25°C | |
4 x 2 x 1 in. (103 x 63 x 33 mm) |
Lithium (TL-5903) | |
mini USB, SD card slot | |
103 mm |